missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serotonin Antibody (YC5/45), Janelia Fluor™ 646, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB100-65037JF646
This item is not returnable.
View return policy
Description
Serotonin Monoclonal antibody specifically detects Serotonin in All samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry (Whole-Mount)
Specifications
| Serotonin | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 5-HT, 5-hydroxytryptamne | |
| Serotonin conjugated to Bovine Serum Albumin. | |
| 0.1 mL | |
| Neuroscience | |
| All species | |
| Purified |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry (Whole-Mount) | |
| YC5/45 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction