missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin A1/alpha 1-Antitrypsin Antibody (OTI9A1) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74095
This item is not returnable.
View return policy
Description
Serpin A1/alpha 1-Antitrypsin Monoclonal specifically detects Serpin A1/alpha 1-Antitrypsin in Human, Canine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, CyTOF-ready.
Specifications
| Serpin A1/alpha 1-Antitrypsin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:500-1000, Flow Cytometry 1:100, Immunohistochemistry 1:150, Immunoprecipitation 2 μg/500 μL, CyTOF-ready | |
| A1A, AATMGC23330, Alpha-1 protease inhibitor, Alpha-1-antiproteinase, alpha-1-antitrypsin, antitrypsin), member 1, member 1, PIMGC9222, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serine (or cysteine) proteinase inhibitor, clade A, member 1, Serpin A1, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin) | |
| Mouse | |
| 46.7 kDa | |
| 100 μg | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, CyTOF | |
| OTI9A1 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| SERPINA1 | |
| Full-length protein expressed in 293T cell transfected with human SERPINA1 expression vector | |
| Affinity Purified | |
| Stem Cell Markers | |
| 5265 | |
| Human, Canine, Primate, Monkey | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction