missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin A12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-15117
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Serpin A12 Polyclonal specifically detects Serpin A12 in Human samples. It is validated for Western Blot.
Especificaciones
| Serpin A12 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| antitrypsin), member 12, member 12, OL-64Visceral adipose-specific serpin, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serpin A12, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), Vaspin, Visceral adipose tissue-derived serine protease inhibitor | |
| Rabbit | |
| 47 kDa | |
| 0.1 mL | |
| metabolism | |
| 145264 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.46mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q8IW75 | |
| SERPINA12 | |
| Recombinant protein encompassing a sequence within the center region of human SERPINA12. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido