missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin A3/alpha 1-Antichymotrypsin Rabbit anti-Human, Clone: 6, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89404-50UL
This item is not returnable.
View return policy
Description
Serpin A3/alpha 1-Antichymotrypsin Monoclonal specifically detects Serpin A3/alpha 1-Antichymotrypsin in Human samples. It is validated for ELISA, Sandwich ELISA Detection.
Specifications
| Serpin A3/alpha 1-Antichymotrypsin | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 6 | |
| ELISA 1:5000-1:10000, Sandwich ELISA Detection 1:1000-1:10000 | |
| AACTMGC88254, ACTGIG25, alpha-1-antichymotrypsin, antitrypsin), member 3, Cell growth-inhibiting gene 24/25 protein, growth-inhibiting protein 24, growth-inhibiting protein 25, member 3, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serine (or cysteine) proteinase inhibitor, clade A, member 3, Serpin A3, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin) | |
| Recombinant Human Serpin A3/alpha 1-Antichymotrypsin Protein | |
| 50 μg | |
| Breast Cancer, Cancer | |
| 12 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction