missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin A6/Cortisol Binding Globulin Antibody (037), Alexa Fluor™ 750, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89739AF750
This item is not returnable.
View return policy
Description
Serpin A6/Cortisol Binding Globulin Monoclonal antibody specifically detects Serpin A6/Cortisol Binding Globulin in Human samples. It is validated for ELISA
Specifications
| Serpin A6/Cortisol Binding Globulin | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 866 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 037 | |
| ELISA | |
| antitrypsin), member 6, CBGcorticosteroid-binding globulin, corticosteroid binding globulin, member 6, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, Serpin A6, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), Transcortin | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Serpin A6/Cortisol Binding Globulin (NP_001747.2; Met 1-Val 405). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction