missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin A7/TBG Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98256-50ul
This item is not returnable.
View return policy
Description
Serpin A7/TBG Polyclonal antibody specifically detects Serpin A7/TBG in Mouse samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications
| Serpin A7/TBG | |
| Polyclonal | |
| ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000 | |
| antitrypsin), member 7, FLJ17820, member 7, Serpin A7, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), T4-binding globulin, TBGclade A (alpha-1 antiproteinase, thyroxin-binding globulin, thyroxine-binding globulin | |
| Produced in rabbits immunized with purified, recombinant Mouse Serpin A7/TBG (Accession#: NP_808588.3; Met 1-Gln 418) | |
| 50 μg | |
| Primary | |
| Mouse | |
| Purified |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 6906 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction