missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin A8/Angiotensinogen Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98259AF700
This item is not returnable.
View return policy
Description
Serpin A8/Angiotensinogen Polyclonal specifically detects Serpin A8/Angiotensinogen in Mouse samples. It is validated for Western Blot, ELISA.
Specifications
| Serpin A8/Angiotensinogen | |
| Polyclonal | |
| Western Blot, ELISA | |
| alpha-1 antiproteinase, antitrypsin, angiotensin II, angiotensinogen, angiotensinogen (serpin peptidase inhibitor, clade A, member 8), ANHU, FLJ92595, FLJ97926, pre-angiotensinogen, serine (or cysteine) proteinase inhibitor, Serpin A8, SERPINA8angiotensin I | |
| Produced in rabbits immunized with purified, recombinant Serpin A8/Angiotensinogen (P11859; Met 1-Val 477). | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 183 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction