missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin B3/SCCA1 Antibody (CPTC-SERPINB3-2), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08536UV
This item is not returnable.
View return policy
Description
Serpin B3/SCCA1 Monoclonal antibody specifically detects Serpin B3/SCCA1 in Human samples. It is validated for Western Blot, Protein Array
Specifications
| Serpin B3/SCCA1 | |
| Monoclonal | |
| DyLight 350 | |
| HsT1196, Protein T4-A, SCC, SCCA, SCCA1SCCA-1, SCCA-PD, serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 3, serpin B3, serpin peptidase inhibitor, clade B (ovalbumin), member 3, Squamous cell carcinoma antigen 1, T4-A | |
| Recombinant human full-length Serpin B3/SCCA1 protein (Uniprot: P29508) | |
| 100 μg | |
| Cancer | |
| 6317 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Protein Array | |
| CPTC-SERPINB3-2 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction