missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin B3/SCCA1 Antibody (OTI3C2), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74108R
This item is not returnable.
View return policy
Description
Serpin B3/SCCA1 Monoclonal specifically detects Serpin B3/SCCA1 in Human, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Serpin B3/SCCA1 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SERPINB3 | |
| Full length recombinant protein of human Serpin B3/SCCA1 (NP_008850) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Rat, Canine, Primate, Monkey | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI3C2 | |
| Western Blot, Immunohistochemistry | |
| HsT1196, Protein T4-A, SCC, SCCA, SCCA1SCCA-1, SCCA-PD, serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 3, serpin B3, serpin peptidase inhibitor, clade B (ovalbumin), member 3, Squamous cell carcinoma antigen 1, T4-A | |
| Mouse | |
| Affinity Purified | |
| Cancer | |
| 6317.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction