Learn More
Serpin B6 Antibody (309), mFluor Violet 450 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90150MFV450
This item is not returnable.
View return policy
Description
Serpin B6 Monoclonal antibody specifically detects Serpin B6 in Human samples. It is validated for ELISA
Specifications
| mFluor Violet 450 SE | |
| Rabbit | |
| RUO | |
| Store at 4C in the dark. |