missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin B6 Antibody (OTI1C5), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74111PCP
This item is not returnable.
View return policy
Description
Serpin B6 Monoclonal specifically detects Serpin B6 in Human, Mouse, Monkey samples. It is validated for Flow Cytometry.
Specifications
| Serpin B6 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| SERPINB6 | |
| Full length recombinant protein of human Serpin B6 (NP_004559) produced in HEK293T cells. | |
| 0.1 mL | |
| 5269.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI1C5 | |
| Flow Cytometry | |
| CAPMGC111370, Cytoplasmic antiproteinase, deafness, autosomal recessive 91, DFNB91, Peptidase inhibitor 6, PI-6, PI6MSTP057, Placental thrombin inhibitor, protease inhibitor 6 (placental thrombin inhibitor), PTIDKFZp686I04222, serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 6, serpin B6, serpin peptidase inhibitor, clade B (ovalbumin), member 6, SPI3 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Monkey | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction