Learn More
Serpin C1/Antithrombin III Mouse, anti-Human, CoraLite Plus 488, Clone: 2B2E5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66052-20UL
Description
The Serpin C1/Antithrombin III antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Serpin C1/Antithrombin III. This antibody recognizes human, mouse, rat antigen. The Serpin C1/Antithrombin III antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| Serpin C1/Antithrombin III | |
| 2B2E5 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat | |
| 155 kDa | |
| IgG1 |
| Immunofluorescence | |
| 1000 μg/mL | |
| Antithrombin, Antithrombin III, AT3, ATIII, HEPARIN COFACTOR I, Serpin C1, SERPINC1 | |
| 20 μL | |
| 462 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |