Learn More
Serpin C1/Antithrombin III Mouse, anti-Human, CoraLite Plus 488, Clone: 2B2E5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-66052-20UL
 View more versions of this product
Product Code. 18037075
  • € 145.00 / 20µL
Description
The Serpin C1/Antithrombin III antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Serpin C1/Antithrombin III. This antibody recognizes human, mouse, rat antigen. The Serpin C1/Antithrombin III antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
Serpin C1/Antithrombin III
2B2E5
CoraLite Plus 488
Mouse
Primary
Human, Mouse, Rat
155 kDa
IgG1
Immunofluorescence
1000 μg/mL
Antithrombin, Antithrombin III, AT3, ATIII, HEPARIN COFACTOR I, Serpin C1, SERPINC1
20 μL
462
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.