missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit IgG Isotype Control, FITC, Novus Biologicals™
Rabbit Antibody
Brand: Novus Biologicals NBP1-96877
This item is not returnable.
View return policy
Description
IgG specifically detects IgG in Rabbit samples. It is validated for SDS-Page.
Specifications
| IgG | |
| Fluorescein | |
| Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide | |
| IGHG1 | |
| Multi-step | |
| RUO | |
| 3500 | |
| Reconstitute with 1.0 ml deionized water (or equivalent). | |
| Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| SDS-Page | |
| SDS-Page 1:100 - 1:2000 | |
| Immunoglobulin G, ImmunoglobulinG | |
| Rabbit | |
| 1 mg | |
| Isotype Control (Primary) | |
| Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Fluorescein, anti-Rabbit IgG and anti-Rabbit Serum. | |
| Rabbit | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction