Learn More
Serpin D1/Heparin Cofactor II Antibody (008), Allophycocyanin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-89392APC
Product Code. 18626450
  • € 607.00 / 0.10mL
Description
Serpin D1/Heparin Cofactor II Monoclonal antibody specifically detects Serpin D1/Heparin Cofactor II in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Serpin D1/Heparin Cofactor II
Monoclonal
APC
PBS
Rabbit
Protein A purified
RUO
Primary
Human
Purified
Immunohistochemistry (Paraffin)
008
Immunohistochemistry-Paraffin
HC2LS2, HCF2clade D (heparin cofactor), member 1, HC-II, Heparin cofactor II, HLS2HCII, leuserpin 2, Protease inhibitor leuserpin-2, Serpin D1, serpin peptidase inhibitor, clade D (heparin cofactor), member 1
This antibody was obtained from a rabbit immunized with purified, recombinant Human Serpin D1/Heparin Cofactor II (Accession#: NP_000176.2; Met1-Ser499).
0.1 mL
Cardiovascular Biology
3053
Store at 4C in the dark.