missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin D1/Heparin Cofactor II Antibody (25), DyLight 488, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90429G
This item is not returnable.
View return policy
Description
Serpin D1/Heparin Cofactor II Monoclonal specifically detects Serpin D1/Heparin Cofactor II in Mouse samples. It is validated for Western Blot, ELISA.
Specifications
| Serpin D1/Heparin Cofactor II | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| No cross-reactivity in ELISA with: Human SerpinA1/ A1AT Human SerpinA3/ AACT Human SerpinA5/ PROCI Mouse SerpinF2/ AAP | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| 25 | |
| Western Blot, ELISA | |
| HC2LS2, HCF2clade D (heparin cofactor), member 1, HC-II, Heparin cofactor II, HLS2HCII, leuserpin 2, Protease inhibitor leuserpin-2, Serpin D1, serpin peptidase inhibitor, clade D (heparin cofactor), member 1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Serpin D1/Heparin Cofactor II (NP_032249.3; Met 1-Ser 478). | |
| 0.1 mL | |
| Cardiovascular Biology | |
| 3053 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction