missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin E1/PAI-1 Antibody (Sanofi patent anti-PAI-1), mFluor Violet 500 SE, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-27963MFV500
This item is not returnable.
View return policy
Description
Serpin E1/PAI-1 Monoclonal antibody specifically detects Serpin E1/PAI-1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Serpin E1/PAI-1 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| Endothelial plasminogen activator inhibitor, PAI1PAI-1, PAISerpin E1, PLANH1plasminogen activator inhibitor 1, serine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 1, serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitortype 1), member 1 | |
| SERPINE1 | |
| 0.1 mL | |
| Cancer | |
| 5054 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Sanofi patent anti-PAI-1 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction