missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin E2/PN1 Antibody (OTI2C9), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74117JF646
This item is not returnable.
View return policy
Description
Serpin E2/PN1 Monoclonal specifically detects Serpin E2/PN1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| Serpin E2/PN1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SERPINE2 | |
| Full length recombinant protein of human Serpin E2/PN1 (NP_006207) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI2C9 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| GDNserine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 2, glia-derived nexin, glial-derived neurite promoting factor, Peptidase inhibitor 7, PI-7, PI7DKFZp686A13110, PN-1, PN1nexin, PNI, Protease nexin 1, Protease nexin I, Serpin E2, serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitortype 1), member 2 | |
| Mouse | |
| Affinity Purified | |
| Neuroscience | |
| 5270.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction