Learn More
Serpin F1/PEDF Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB600-1493
Product Code. 18776203
  • € 493.00 / 0.10mg
Description
Serpin F1/PEDF Polyclonal specifically detects Serpin F1/PEDF in Human, Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyclonal
Store at -20C. Avoid freeze-thaw cycles.