missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin G1/C1 Inhibitor Antibody (18), HRP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89738H
This item is not returnable.
View return policy
Description
Serpin G1/C1 Inhibitor Monoclonal specifically detects Serpin G1/C1 Inhibitor in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA Detection.
Specifications
| Serpin G1/C1 Inhibitor | |
| Monoclonal | |
| HRP | |
| C1IN, C1INH, C1NH, HAE1, HAE2, SERPING1 serpin peptidase inhibitor, clade G (C1 inhibitor), member 1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Serpin G1/C1 Inhibitor (NP_000053.2; Met 1-Ala 500). | |
| 0.1 mL | |
| Primary | |
| No cross-reactivity in ELISA with: Human SerpinA1/ A1AT Human SerpinA3/ SERPINA3/ AACT Human SerpinA5/ PROCI Human SerpinC1/SERPINC1/ Antithrombin-III Human SerpinD1/ SERPIND1/ HC2 Human SerpinF1/ SERPINF1/ PEDF Human SerpinF2/ SERPINF2 Human SerpinI1/ SERPINI1/ Protease inhibitor 12 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| 18 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 710 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction