missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin G1/C1 Inhibitor Rabbit anti-Human, Clone: 18, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 379.00 - € 552.00
Specifications
| Antigen | Serpin G1/C1 Inhibitor |
|---|---|
| Clone | 18 |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 0.5-2 ul/mg of lysate, Sandwich ELISA Detection 1:1000-1:10000 |
| Applications | Western Blot, ELISA, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18224566
|
Novus Biologicals
NBP2-89738-50UL |
50 μg |
€ 379.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18276958
|
Novus Biologicals
NBP2-89738-100UL |
100 μg |
€ 552.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Serpin G1/C1 Inhibitor Monoclonal specifically detects Serpin G1/C1 Inhibitor in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Sandwich ELISA Detection.Specifications
| Serpin G1/C1 Inhibitor | |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 0.5-2 ul/mg of lysate, Sandwich ELISA Detection 1:1000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 710 | |
| IgG | |
| Protein A purified |
| 18 | |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human | |
| C1IN, C1INH, C1NH, HAE1, HAE2, SERPING1 serpin peptidase inhibitor, clade G (C1 inhibitor), member 1 | |
| Recombinant Human Serpin G1/C1 Inhibitor protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title