missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin I2 Antibody (018), mFluor Violet 500 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89740MFV500
This item is not returnable.
View return policy
Description
Serpin I2 Monoclonal antibody specifically detects Serpin I2 in Human samples. It is validated for Flow Cytometry
Specifications
| Serpin I2 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 5276 | |
| Store at 4C in the dark. | |
| IgG |
| Flow Cytometry | |
| 018 | |
| Flow Cytometry | |
| MEPIpancpin, Myoepithelium-derived serine protease inhibitor, Pancpin, Pancreas-specific protein TSA2004, Peptidase inhibitor 14, PI-14, PI14clade I (neuroserpin), member 2, protease inhibitor 14, serine (or cysteine) proteinase inhibitor, clade I (pancpin), member 2, serpin I2, serpin peptidase inhibitor, clade I (pancpin), member 2, SERPIN12 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Serpin I2 (NP_006208.1; Met 1-Leu 405). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction