Learn More
Invitrogen™ SERPINA11 Recombinant Rabbit Monoclonal Antibody (202)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA531232
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Mouse SerpinA11.
The protein belongs to the serpin family. SERPINA11 has serine-type endopeptidase inhibitor activity, negative regulation of endopeptidase activity, regulation of proteolysis, serine-type endopeptidase inhibitor activity.
Specifications
| SERPINA11 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q8CIE0 | |
| SERPINA11 | |
| Recombinant Mouse SerpinA11 Protein (Met1-Gly424). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| ELISA | |
| 202 | |
| Unconjugated | |
| SERPINA11 | |
| antiproteinase-like 2; Gm895; serine (or cysteine) peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 11; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 11; serine (or cysteine) proteinase inhibitor, clade A, member 11; serpin A11; serpin family A member 11; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 11; Serpina11 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 380780 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.