Learn More
Invitrogen™ SERPINA3 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA532260
 View more versions of this product
Product Code. 13496777
  • € 306.00 / 500µL
Description
A suggested positive control is tonsil tissue.
SERPINA encodes Alpha 1-ACT. It is an early-stage acute-phase plasma protein and a serpin that preferentially inactivates chymotrypsin, cathepsin G, and chymase. Alpha-1-ACT, a serine protease inhibitor, is tightly associated with amyloid plaques in Alzheimer's disease (AD) and in normal aged human and monkey brain. Regulation of the serine proteases and serine protease inhibitors plays an important role in neuromuscular differentiation. Prostate specific antigen (PSA), a chymotrypsin-like serine protease, is predominantly complexed to Alpha-1-ACT.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SERPINA3
Polyclonal
PBS with 1% BSA and 0.1% sodium azide; pH 7.6
P01011
SERPINA3
Purified human alpha-1-antichymotrypsin.
500 μL
Primary
Human
Antibody
IgG
Immunohistochemistry (Paraffin)
Unconjugated
SERPINA3
AACT; ACT; alpha-1-antichymotrypsin; Alpha-1-antichymotrypsin His-Pro-less; Cell growth-inhibiting gene 24/25 protein; GIG24; GIG25; Growth inhibiting protein 24; Growth inhibiting protein 25; growth-inhibiting protein 24; growth-inhibiting protein 25; MGC88254; Serine (or cysteine) proteinase inhibitor clade A member 3; serine (or cysteine) proteinase inhibitor, clade A, member 3; serpin A3; serpin family A member 3; Serpin peptidase inhibitor clade A (alpha 1 antiproteinase antitrypsin) member 3; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 3; SERPINA3
Rabbit
Antigen affinity chromatography
RUO
12
-20°C, Avoid Freeze/Thaw Cycles