missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpina3k Rabbit anti-Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55480-1-AP-20UL
This item is not returnable.
View return policy
Specifications
| Serpina3k | |
| Polyclonal | |
| Unconjugated | |
| Serpina3k | |
| 1300001I07Rik, contrapsin, D12Rp54, MMCM2, MMSpi2, RP54, Serpina3k, Spi 2, Spi2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20714 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.67 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P07759 | |
| Serpina3k | |
| Peptide | |
| 20 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction