missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERPINB1 Antibody (04), HRP , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-05846H
This item is not returnable.
View return policy
Description
SERPINB1 Monoclonal antibody specifically detects SERPINB1 in Human samples. It is validated for ELISA
Specifications
| SERPINB1 | |
| Monoclonal | |
| HRP | |
| EILEI, ELANH2MNEI, elastase, leukocyte elastase inhibitor, Monocyte/neutrophil elastase inhibitor, Peptidase inhibitor 2, PI-2, PI2M/NEI, protease inhibitor 2 (elastase), monocyte/neutrophil derived, serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 1, Serpin B1, serpin peptidase inhibitor, clade B (ovalbumin), member 1 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SERPINB1 (P30740; Met 1-Pro 379). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 04 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 1992 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction