missing translation for 'onlineSavingsMsg'
Learn More

SERPINB1 Monoclonal Antibody (OTI2D11), TrueMAB™, OriGene

Product Code. p-200015615
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17577562 30 μL 30µL
17557562 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17577562 Supplier Origene Technologies Supplier No. TA800094S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SERPINB1 Monoclonal antibody specifically detects SERPINB1 in Canine, Human, Rat samples. It is validated for Western Blot

Specifications

Antigen SERPINB1
Applications Western Blot
Classification Monoclonal
Clone OTI2D11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SERPINB1
Gene Accession No. P30740, Q4G075
Gene Alias 1190005M04Rik; AI325983; Anti elastase; anti-elastase; EGK_14416; EI; EIA; ELANH2; epididymis luminal protein 57; epididymis secretory protein Li 27; HEL57; HEL-S-27; L M/N MN PI2; LEI; leukocyte elastase inhibitor; Leukocyte elastase inhibitor A; M/NEI; MNEI; Monocyte/neutrophil elastase inhibitor; ovalbumin; peptidase inhibitor 2; PI2; PI-2; protease inhibitor 2 (anti-elastase), monocyte/neutrophil derived; serine (or cysteine) peptidase inhibitor, clade B, member 1a; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 1; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 1a; serine (or cysteine) proteinase inhibitor, clade B, member 1a; serine protease inhibitor EIA; serpin; Serpin B1; Serpin B1a; serpin family B member 1; serpin peptidase inhibitor, clade B (ovalbumin), member 1; Serpinb1; Serpinb1a
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 77-306 of human SERPINB1 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1992, 291091, 488191
Target Species Canine, Human, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.