missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SERPINB1 Polyclonal Antibody

Rabbit Polyclonal Antibody
€ 208.00 - € 476.00
Specifications
| Antigen | SERPINB1 |
|---|---|
| Concentration | 0.5 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17730664
![]() |
Invitrogen™
PA5119256 |
200 μL |
€ 476.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17763014
|
Invitrogen™
PA5121618 |
20 μL |
€ 208.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SERPINB1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Regulates the activity of the neutrophil proteases elastase, cathepsin G, proteinase-3, chymase, chymotrypsin, and kallikrein-3.Specifications
| SERPINB1 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse | |
| SERPINB1 | |
| 1190005M04Rik; AI325983; Anti elastase; anti-elastase; EGK_14416; EI; EIA; ELANH2; epididymis luminal protein 57; epididymis secretory protein Li 27; HEL57; HEL-S-27; L M/N MN PI2; LEI; leukocyte elastase inhibitor; Leukocyte elastase inhibitor A; M/NEI; MNEI; Monocyte/neutrophil elastase inhibitor; ovalbumin; peptidase inhibitor 2; PI2; PI-2; protease inhibitor 2 (anti-elastase), monocyte/neutrophil derived; serine (or cysteine) peptidase inhibitor, clade B, member 1a; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 1; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 1a; serine (or cysteine) proteinase inhibitor, clade B, member 1a; serine protease inhibitor EIA; serpin; Serpin B1; Serpin B1a; serpin family B member 1; serpin peptidase inhibitor, clade B (ovalbumin), member 1; Serpinb1; Serpinb1a | |
| SERPINB1, Serpinb1a | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P30740, Q9D154 | |
| 1992, 66222 | |
| Recombinant protein Leukocyte Elastase Inhibitor. The antigen corresponds to amino acid range 184-368 of the target protein. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title