missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SERPINB1 Polyclonal Antibody

Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Regulates the activity of the neutrophil proteases elastase, cathepsin G, proteinase-3, chymase, chymotrypsin, and kallikrein-3.
Specifications
Specifications
| Antigen | SERPINB1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SERPINB1 |
| Gene Accession No. | P30740, Q4G075, Q9D154 |
| Gene Alias | 1190005M04Rik; AI325983; Anti elastase; anti-elastase; EGK_14416; EI; EIA; ELANH2; epididymis luminal protein 57; epididymis secretory protein Li 27; HEL57; HEL-S-27; L M/N MN PI2; LEI; leukocyte elastase inhibitor; Leukocyte elastase inhibitor A; M/NEI; MNEI; Monocyte/neutrophil elastase inhibitor; ovalbumin; peptidase inhibitor 2; PI2; PI-2; protease inhibitor 2 (anti-elastase), monocyte/neutrophil derived; serine (or cysteine) peptidase inhibitor, clade B, member 1a; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 1; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 1a; serine (or cysteine) proteinase inhibitor, clade B, member 1a; serine protease inhibitor EIA; serpin; Serpin B1; Serpin B1a; serpin family B member 1; serpin peptidase inhibitor, clade B (ovalbumin), member 1; Serpinb1; Serpinb1a |
| Gene Symbols | SERPINB1, Serpinb1a |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?