missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERPINB10 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24382MFV500
This item is not returnable.
View return policy
Description
SERPINB10 Polyclonal antibody specifically detects SERPINB10 in Human samples. It is validated for Western Blot
Specifications
| SERPINB10 | |
| Polyclonal | |
| Western Blot | |
| bomapin, Peptidase inhibitor 10, PI-10, PI10protease inhibitor 10 (ovalbumin type, bomapin), serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 10, serpin B10, serpin peptidase inhibitor, clade B (ovalbumin), member 10 | |
| Reacts with residues 316-330 [SQSKADFSGMSSARN] of the human SERPINB10 protein. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 5273 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction