missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERPINB10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 216.00 - € 307.65
Specifications
| Antigen | SERPINB10 |
|---|---|
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18629651
|
Novus Biologicals
NBP2-98324-100ul |
100 μg |
€ 326.00 € 307.65 / 100µL Save € 18.35 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18683875
|
Novus Biologicals
NBP2-98324-50ul |
50 μg |
€ 216.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SERPINB10 Polyclonal antibody specifically detects SERPINB10 in Mouse samples. It is validated for ELISASpecifications
| SERPINB10 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| bomapin, Peptidase inhibitor 10, PI-10, PI10protease inhibitor 10 (ovalbumin type, bomapin), serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 10, serpin B10, serpin peptidase inhibitor, clade B (ovalbumin), member 10 | |
| Produced in rabbits immunized with purified, recombinant Mouse SERPINB10 (Uniprot#: Q8K1K6-1; Met1-Pro397) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| ELISA 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 5273 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.