missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERPINB12 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-93572AF488
This item is not returnable.
View return policy
Description
SERPINB12 Polyclonal antibody specifically detects SERPINB12 in Human samples. It is validated for Western Blot
Specifications
| SERPINB12 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 89777 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 488 | |
| MGC119247, MGC119248, serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 12, serpin B12, serpin peptidase inhibitor, clade B (ovalbumin), member 12, YUKOPIN | |
| Reacts with residues 260-275 [SHSKDNLKGLEELERK] of the human SERPINB12 protein (NM_080474) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction