Learn More
SERPINB12 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB100-93572APC
Product Code. 18564294
  • € 508.00 / 0.10mL
Description
SERPINB12 Polyclonal antibody specifically detects SERPINB12 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyclonal
MGC119247, MGC119248, serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 12, serpin B12, serpin peptidase inhibitor, clade B (ovalbumin), member 12, YUKOPIN
Reacts with residues 260-275 [SHSKDNLKGLEELERK] of the human SERPINB12 protein (NM_080474)
Store at 4C in the dark.