missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERPINB13 Antibody (OTI2B6), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74126AF350
This item is not returnable.
View return policy
Description
SERPINB13 Monoclonal specifically detects SERPINB13 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| SERPINB13 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| SERPINB13 | |
| Full length recombinant protein of human SERPINB13 (NP_036529) produced in HEK293T cells. | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI2B6 | |
| Western Blot, Immunohistochemistry | |
| HaCaT UV-repressible serpin, Headpin, HSHUR7SEQ, HUR7, HURPIN, MGC126870, Peptidase inhibitor 13, PI-13, PI13hurpin, protease inhibitor 13 (hurpin, headpin), Proteinase inhibitor 13, serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 13, serpin B13, serpin peptidase inhibitor, clade B (ovalbumin), member 13, UV-B repressed sequence, HUR 7 | |
| Mouse | |
| 0.1 mL | |
| 5275 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction