missing translation for 'onlineSavingsMsg'
Learn More

SERPINB13 Monoclonal Antibody (OTI1B12), TrueMAB™, OriGene

Product Code. p-200011617
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17539801 30 μL 30µL
17519801 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17539801 Supplier Origene Technologies Supplier No. TA503674S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SERPINB13 Monoclonal antibody specifically detects SERPINB13 in Human samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen SERPINB13
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI1B12
Concentration 0.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SERPINB13
Gene Accession No. Q9UIV8
Gene Alias 5430417G24; HaCaT UV-repressible serpin; headpin; HSHUR7SEQ; HUR7; HURPIN; Peptidase inhibitor 13; PI13; PI-13; protease inhibitor 13 (hurpin, headpin); proteinase inhibitor 13; serine (or cysteine) peptidase inhibitor, clade B (ovalbumin), member 13; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 13; serpin B13; serpin family B member 13; serpin peptidase inhibitor, clade B (ovalbumin), member 13; SERPINB13; UV-B repressed sequence, HUR 7
Host Species Mouse
Immunogen Full length human recombinant protein of human SERPINB13 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5275
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.