Learn More
Invitrogen™ SERPINB13 Monoclonal Antibody (OTI2B6)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA525718
Product Code. 15985314
  • € 572.00 / 100µL
Description
SERPINB13 Monoclonal Antibody for Western Blot, IHC (P)
Members of the human serpin family regulate a diverse array of serine and cysteine proteinases associated with essential biological processes such as fibrinolysis, coagulation, inflammation, cell mobility, cellular differentiation, and apoptosis. Most serpins are secreted and attain physiologic concentrations in the blood and extracellular fluids. However, a subset of the serpin superfamily, the ov-serpins, also resides intracellularly. Differential display was used to identify a novel serpin (headpin or SERPINB13) that is under expressed in squamous cell cancers of the oral cavity. SERPINB13 is expressed in normal oral mucosal tissue, skin, and cultured keratinocytes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SERPINB13
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9UIV8
SERPINB13
Full length human recombinant protein of SERPINB13 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI2B6
Unconjugated
SERPINB13
5430417G24; HaCaT UV-repressible serpin; headpin; HSHUR7SEQ; HUR7; HURPIN; Peptidase inhibitor 13; PI13; PI-13; protease inhibitor 13 (hurpin, headpin); proteinase inhibitor 13; serine (or cysteine) peptidase inhibitor, clade B (ovalbumin), member 13; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 13; serpin B13; serpin family B member 13; serpin peptidase inhibitor, clade B (ovalbumin), member 13; SERPINB13; UV-B repressed sequence, HUR 7
Mouse
Affinity Chromatography
RUO
5275
-20°C, Avoid Freeze/Thaw Cycles