Learn More
SERPINB13 Mouse anti-Human, Clone: OTI2B6, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF503672
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Members of the human serpin family regulate a diverse array of serine and cysteine proteinases associated with essential biological processes such as fibrinolysis, coagulation, inflammation, cell mobility, cellular differentiation, and apoptosis. Most serpins are secreted and attain physiologic concentrations in the blood and extracellular fluids. However, a subset of the serpin superfamily, the ov-serpins, also resides intracellularly. Differential display was used to identify a novel serpin (headpin or SERPINB13) that is underexpressed in squamous cell cancers of the oral cavity. SERPINB13 is expressed in normal oral mucosal tissue, skin, and cultured keratinocytes.Specifications
| SERPINB13 | |
| Monoclonal | |
| Unconjugated | |
| SERPINB13 | |
| HSHUR7SEQ, HUR7, PI13, headpin | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5275 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2B6 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9UIV8 | |
| SERPINB13 | |
| Full length human recombit protein of human SERPINB13 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.