missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SERPINB13 Polyclonal Antibody

Description
Immunogen sequence: MDSLGAVSTR LGFDLFKELK KTNDGNIFFS PVGILTAIGM VLLGTRGATA SQLEEVFHSE KETKSSRIKA EEKEVIENTE AVHQQFQKFL TEISKLTNDY ELNITNRLFG EKTYLFLQKY LDYVEKYYHA SLEPVDFVNA; Positive Samples: Mouse uterus, Rat uterus; Cellular Location: Cytoplasm.
Members of the human serpin family regulate a diverse array of serine and cysteine proteinases associated with essential biological processes such as fibrinolysis, coagulation, inflammation, cell mobility, cellular differentiation, and apoptosis. Most serpins are secreted and attain physiologic concentrations in the blood and extracellular fluids. However, a subset of the serpin superfamily, the ov-serpins, also resides intracellularly. Differential display was used to identify a novel serpin (headpin or SERPINB13) that is under expressed in squamous cell cancers of the oral cavity. SERPINB13 is expressed in normal oral mucosal tissue, skin, and cultured keratinocytes.
Specifications
Specifications
| Antigen | SERPINB13 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.40 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | SERPINB13 |
| Gene Accession No. | Q8CDC0, Q9UIV8 |
| Gene Alias | 5430417G24; HaCaT UV-repressible serpin; headpin; HSHUR7SEQ; HUR7; HURPIN; Peptidase inhibitor 13; PI13; PI-13; protease inhibitor 13 (hurpin, headpin); proteinase inhibitor 13; serine (or cysteine) peptidase inhibitor, clade B (ovalbumin), member 13; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 13; serpin B13; serpin family B member 13; serpin peptidase inhibitor, clade B (ovalbumin), member 13; SERPINB13; UV-B repressed sequence, HUR 7 |
| Gene Symbols | SERPINB13 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?