missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SerpinB2 Antibody (930109) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8550JF525
This item is not returnable.
View return policy
Description
SerpinB2 Monoclonal antibody specifically detects SerpinB2 in Human samples. It is validated for Flow Cytometry,Western Blot,Immunocytochemistry,CyTOF,Gene Knock-Out
Specifications
| SerpinB2 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| HsT1201, Monocyte Arg-serpin, PAI, PAI2Urokinase inhibitor, Placental plasminogen activator inhibitor, PLANH2PAI-2, plasminogen activator inhibitor 2, plasminogen activator inhibitor, type II (arginine-serpin), serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 2, Serpin B2, serpin peptidase inhibitor, clade B (ovalbumin), member 2 | |
| E. coli-derived recombinant human Serpin B2, Glu2-Ala65, Ala99-Pro415, Accession # P05120 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry, CyTOF, Gene Knock-Out | |
| 930109 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Apoptosis, Cancer | |
| 5055 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction