missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SerpinB2 Antibody (OTI1G3), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74131PEATT594
This item is not returnable.
View return policy
Description
SerpinB2 Monoclonal specifically detects SerpinB2 in Human samples. It is validated for Flow Cytometry.
Specifications
| SerpinB2 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| SERPINB2 | |
| Full length recombinant protein of human SerpinB2 (NP_002566) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| OTI1G3 | |
| Flow Cytometry | |
| HsT1201, Monocyte Arg-serpin, PAI, PAI2Urokinase inhibitor, Placental plasminogen activator inhibitor, PLANH2PAI-2, plasminogen activator inhibitor 2, plasminogen activator inhibitor, type II (arginine-serpin), serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 2, Serpin B2, serpin peptidase inhibitor, clade B (ovalbumin), member 2 | |
| Mouse | |
| Affinity Purified | |
| Apoptosis, Cancer | |
| 5055.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction