missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SERPINB3 Monoclonal Antibody (D5), Invitrogen™

Mouse Monoclonal Antibody
€ 185.00 - € 415.00
Specifications
| Antigen | SERPINB3 |
|---|---|
| Clone | D5 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17909532
|
Invitrogen™
MA551076 |
20 μL |
€ 185.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17980443
![]() |
Invitrogen™
MA551075 |
200 μL |
€ 415.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
May act as a papain-like cysteine protease inhibitor to modulate the host immune response against tumor cells. Also functions as an inhibitor of UV-induced apoptosis via suppression of the activity of c-Jun NH(2)-terminal kinase (JNK1). [UniProt]Specifications
| SERPINB3 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| HsT1196; LOC483954; Protein T4-A; SCC; SCCA; SCCA1; SCCA-1; SCCA-PD; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 3; serine protease inhibitor B3; serpin B3; serpin family B member 3; serpin peptidase inhibitor, clade B (ovalbumin), member 3; serpin peptidase inhibitor, clade B (ovalbumin), member 4; SERPINB3; SERPINB4; spb3; Squamous cell carcinoma antigen 1; SSCA1; T4-A | |
| SERPINB3 | |
| IgG1 κ | |
| Protein A/G | |
| SERPINB3 |
| D5 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P29508 | |
| 6317 | |
| Recombinant Human SERPINB3 protein, Arg144-Thr298 (Accession #Q9BYH9). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title