missing translation for 'onlineSavingsMsg'
Learn More

SERPINB3 Monoclonal Antibody (OTI1A10), TrueMAB™, OriGene

Product Code. p-200013639
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17556522 30 μL 30µL
17536522 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17556522 Supplier Origene Technologies Supplier No. TA506888S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SERPINB3 Monoclonal antibody specifically detects SERPINB3 in Human samples. It is validated for Western Blot

Specifications

Antigen SERPINB3
Applications Western Blot
Classification Monoclonal
Clone OTI1A10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SERPINB3
Gene Accession No. P29508
Gene Alias HsT1196; LOC483954; Protein T4-A; SCC; SCCA; SCCA1; SCCA-1; SCCA-PD; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 3; serine protease inhibitor B3; serpin B3; serpin family B member 3; serpin peptidase inhibitor, clade B (ovalbumin), member 3; serpin peptidase inhibitor, clade B (ovalbumin), member 4; SERPINB3; SERPINB4; spb3; Squamous cell carcinoma antigen 1; SSCA1; T4-A
Host Species Mouse
Immunogen Full length human recombinant protein of human SERPINB3 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6317
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.