missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SERPINB9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119075
This item is not returnable.
View return policy
Description
SERPINB9 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
PI9 belongs to the large superfamily of serine proteinase inhibitors (serpins), which bind to and inactivate serine proteinases. These interactions are involved in many cellular processes, including coagulation, fibrinolysis, complement fixation, matrix remodeling, and apoptosis (Sprecher et al., 1995 [PubMed 8530382]).
Specifications
| SERPINB9 | |
| Polyclonal | |
| Unconjugated | |
| SERPINB9 | |
| BB283241; CAP3; CAP-3; Cytoplasmic antiproteinase 3; ovalbumin; peptidase inhibitor 9; PI9; PI-9; protease inhibitor 9 (ovalbumin type); serine (or cysteine) peptidase inhibitor, clade B, member 9; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 9; serine (or cysteine) proteinase inhibitor, clade B, member 9; serine protease inhibitor 6; serpin B9; serpin family B member 9; serpin peptidase inhibitor, clade B (ovalbumin), member 9; serpin peptidase inhibitor, clade B, member 9; SERPINB9; Spi6; testicular tissue protein Li 180 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 20723, 5272 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P50453 | |
| SERPINB9 | |
| Recombinant protein Cytoplasmic Antiproteinase 3. The antigen corresponds to amino acid range 1-376 of the target protein. | |
| 200 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction