missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERPINC1 Mouse anti-Human, Clone: OTI8D5, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA500455
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a plasma protease inhibitor and a member of the serpin superfamily. This protein inhibits thrombin as well as other activated serine proteases of the coagulation system, and it regulates the blood coagulation cascade. The protein includes two functional domains: the heparin binding-domain at the N-terminus of the mature protein, and the reactive site domain at the C-terminus. The inhibitory activity is enhanced by the presence of heparin. More than 120 mutations have been identified for this gene, many of which are known to cause antithrombin-III deficiency.Specifications
| SERPINC1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P01008 | |
| SERPINC1 | |
| Full length human recombit protein of human SERPINC1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry | |
| OTI8D5 | |
| Unconjugated | |
| SERPINC1 | |
| AT3, AT3D, ATIII, THPH7 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 462 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction