missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERPINE2 Monoclonal Antibody (OTI4E4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504596
This item is not returnable.
View return policy
Description
SERPINE2 encodes a member of the serpin family of proteins, a group of proteins that inhibit serine proteases. Thrombin, urokinase, plasmin and trypsin are among the proteases that this family member can inhibit. This gene is a susceptibility gene for chronic obstructive pulmonary disease and for emphysema. Alternative splicing results in multiple transcript variants.Specifications
| SERPINE2 | |
| Monoclonal | |
| 4.02 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P07093, Q07235 | |
| Serpine2 | |
| Full length human recombit protein of human SERPINE2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Canine, Human, Mouse | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI4E4 | |
| Unconjugated | |
| SERPINE2 | |
| GDN, GDNPF, PI-7, PI7, PN-1, PN1, PNI | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 20720, 5270, 608930 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction