missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERPINE2 Monoclonal Antibody (OTI4E4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504596S
This item is not returnable.
View return policy
Description
SERPINE2 Monoclonal antibody specifically detects SERPINE2 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| SERPINE2 | |
| Monoclonal | |
| 4.02 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q14802 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5349 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI4E4 | |
| Unconjugated | |
| FXYD3 | |
| B230326M24Rik; EGK_04851; GDN; GDNPF; Gdnpn1; Glia derived nexin; glia-derived nexin; Glia-derived nexin/protease nexin I; Glia-derived nexin-like protein; glial-derived neurite promoting factor; P17; PAI-1; Peptidase inhibitor 7; Pi7; PI-7; plasminogen activator inhibitor type 1; PN1; Pn-1; PNI; Protease nexin 1; protease nexin I; protease nexin PN-1; protease nexin-1; serine (or cysteine) peptidase inhibitor, clade E, member 2; serine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 2; serine (or cysteine) proteinase inhibitor, clade E, member 2; serine protease inhibitor 4; serine protease-inhibitor 4; Serpin E2; serpin family E member 2; serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 2; serpin peptidase inhibitor, clade E, member 2; SERPINE 2; SERPINE1; Serpine2; Spi4; Spin4 | |
| Full length human recombinant protein of human FXYD3 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction