missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIPK3 Mouse anti-Human, Clone: OTI2B10, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803093S
This item is not returnable.
View return policy
Description
RIP3 is a novel member of RIP Ser/Thr kinase family. It is a potent inducer of apoptosis and is an important component of TNFR1 signaling complex. RIP3 consists of an N-terminal RIP-like-Kinase domain and a unique, non-homologous C-terminus responsible for caspase activation and apoptosis induction. RIP3 is expressed in multiple tissues including hematopoietic cells and plays a functional role in the regulation of apoptosis and NF-kB signaling. It is recruited to the TNFR1 signaling complex in a RIP-dependant manner where it induces apoptosis by activating caspases and/or inhibiting TNFR1-induced NF-kB activation. RIP3 negatively regulates the TLR3-Trif-mediated NF-kB signaling pathway by competing with the binding of Trif to RIP1. The human RIP3 gene is localized in the chromosomal region 14q11.2.Specifications
| RIPK3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y572 | |
| RIPK3 | |
| Human recombit protein fragment corresponding to amino acids 233-518 of human RIPK3 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2B10 | |
| Unconjugated | |
| RIPK3 | |
| RIP3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 11035 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction