missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SERPINE3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562514
This item is not returnable.
View return policy
Description
Immunogen sequence: VLELPYLGSA VSLFLVLPRD KDTPLSHIEP HLTASTIHLW TTSLRRARMD VFLPRFRIQN QFNLKSILNS WGVTDLFDPL KANLKGI Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 83%.
Specifications
| SERPINE3 | |
| Polyclonal | |
| Unconjugated | |
| SERPINE3 | |
| nexin-related serine protease inhibitor; serpin E3; serpin family E member 3; serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 3; SERPINE3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 647174 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A8MV23 | |
| SERPINE3 | |
| Recombinant protein corresponding to Human SERPINE3. Recombinant protein control fragment (Product #RP-98016). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction