missing translation for 'onlineSavingsMsg'
Learn More

SERPING1/C1 Inactivator Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Product Code. 16889863 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16889863 150 μL 150µL
16899863 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16889863 Supplier Proteintech Supplier No. 122591AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene encodes a highly glycosylated plasma protein involved in the regulation of the complement cascade. Its protein inhibits activated C1r and C1s of the first complement component and thus regulates complement activation. Deficiency of this protein is associated with hereditary angioneurotic oedema (HANE). Alternative splicing results in multiple transcript variants encoding the same isoform.

Specifications

Antigen SERPING1/C1 Inactivator
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.19 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene SERPINC1
Gene Accession No. P01008, P05155, P32261, P97290
Gene Alias C1 esterase inhibitor, C1 Inh, C1 inhibiting factor, C1IN, C1INH, C1NH, HAE1, HAE2, Plasma protease C1 inhibitor, Serpin G1, SERPING1
Gene Symbols SERPINC1, SERPING1
Host Species Rabbit
Immunogen SERPING1/C1 Inactivator Fusion Protein Ag2819
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11905, 12258, 462, 710
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.