missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Serratia marcescens Monoclonal Antibody (B/N4N)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA546567
This item is not returnable.
View return policy
Description
Serratia marcescens Monoclonal Antibody for ICC/IF, ELISA
Serratia marcescens (Sma) is a Gram-negative bacterium that is now recognized as an important opportunistic pathogen. It causes a range of nosocomial infections, such as pneumonia, urinary tract infections, bacteremia, meningitis and endocarditis, particularly in immunocompromised patients [PMID: 35315751].
Specifications
| Serratia marcescens | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 10mM sodium azide | |
| Mouse | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG1 |
| ELISA, Immunocytochemistry | |
| B/N4N | |
| Unconjugated | |
| S. marcescens | |
| Native Serratia marcescens (ATCC 13880). | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction