missing translation for 'onlineSavingsMsg'
Learn More

Sertindole, TRC

Product Code. 30011478
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
50 mg
Unit Size:
10mg
2.5mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011478 10 mg 10mg
30007752 50 mg 50mg
30007458 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011478 Supplier Toronto Research Chemicals Supplier No. TRCS27950010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106516-24-9
Molecular Formula C24 H26 Cl F N4 O
Molecular Weight (g/mol) 440.94
IUPAC Name 1-[2-[4-[5-chloro-1-(4-fluorophenyl)indol-3-yl]piperidin-1-yl]ethyl]imidazolidin-2-one
SMILES Fc1ccc(cc1)n2cc(C3CCN(CCN4CCNC4=O)CC3)c5cc(Cl)ccc25

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C24H26ClFN4O/c25-18-1-6-23-21(15-18)22(16-30(23)20-4-2-19(26)3-5-20)17-7-10-28(11-8-17)13-14-29-12-9-27-24(29)31/h1-6,15-17H,7-14H2,(H,27,31)
Formula Weight 440.1779
Chemical Name or Material Sertindole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.